missing translation for 'onlineSavingsMsg'
Learn More

Bis(dibenzylideneacetone)palladium

CAS: 32005-36-0 | C34H28O2Pd | 575.02 g/mol

$156.54 - $156.54

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Molecular Weight (g/mol) 575.02
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC291970010
SDS View Documents
Thermo Scientific Chemicals
291970010
1 g Glass bottle
Each for $156.54
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32005-36-0
575.02
UKSZBOKPHAQOMP-SVLSSHOZSA-N
6505921
[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
C34H28O2Pd
MFCD00051942
bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium

Specifications

32005-36-0
Black to Brown-Red
C34H28O2Pd
MFCD00051942
15,302
Solubility in water: insoluble.
[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
575.02
575.02
Fine Powder
150.0°C
Glass bottle
Pd(C6H5CH=CHC(O)CH=CHC6H5)2
1 g
bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
UKSZBOKPHAQOMP-SVLSSHOZSA-N
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
6505921
16 to 21% (Pd)
Bis(dibenzylideneacetone)palladium
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only