missing translation for 'onlineSavingsMsg'
Learn More

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, 95%

Catalog No. AC447890010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447890010 1 g Glass bottle
AC447890050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447890010 Supplier Thermo Scientific Chemicals Supplier No. 447890010

CAS: 887919-35-9 | C32H56Cl2N2P2Pd | 708.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 887919-35-9
Molecular Formula C32H56Cl2N2P2Pd
Molecular Weight (g/mol) 708.08
MDL Number MFCD09265123
InChI Key DWOZNANUEDYIOF-UHFFFAOYSA-L
Synonym bis di-tert-butyl 4-dimethylaminophenyl phosphine dichloropalladium ii, pd amphos cl2, pdcl2 amphos 2, a-taphos 2pdcl2, dichlorobis di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii, alpha-taphos 2pdcl2, bis 4-di-tert-butylphosphino-n,n-dimethylbenzenamine palladium dichloride
PubChem CID 11714597
IUPAC Name 4-ditert-butylphosphanyl-N,N-dimethylaniline;dichloropalladium
SMILES Cl[Pd]Cl.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C

Specifications

Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 708.07
Percent Purity 95%
Chemical Name or Material Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only