missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, 97%

Catalog No. p-68064
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215760100 10 g Glass bottle
AC215760500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC215760100 Supplier Thermo Scientific Chemicals Supplier No. 215760100
Only null left

CAS: 1271-19-8 | C10H10Cl2Ti | 249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti
Molecular Weight (g/mol) 249
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Titanocene dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 287.0°C to 289.0°C
Color Bordeaux to Red
Density 1.6000g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 10 g
Fieser 06,48
Specific Gravity 1.6
Merck Index 15, 9638
Solubility Information Solubility in water: hydrolyses. Other solubilities: soluble in aromatic hydrocarbons, thf, halogenated, solvents, acetonitrile,, slightly soluble in chloroform, ethanol < 1mg/l,, acetone < 1mg/l
Formula Weight 249
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Bis(cyclopentadienyl)titanium dichloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal
EINECSNumber 215-035-9
RTECSNumber XR2050000
TSCA TSCA

RUO – Research Use Only