missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)nickel, dry

Catalog No. p-7080387
Change view
Click to view available options
Quantity:
2 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8789904 2 g
AA8789918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8789904 Supplier Thermo Scientific Chemicals Supplier No. 08789904
Only null left

CAS: 1271-28-9 | C10H10Ni | 188.883 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 1271-28-9
Molecular Formula C10H10Ni
Molecular Weight (g/mol) 188.883
MDL Number MFCD00001441
InChI Key RQKPFSQDBAZFJV-UHFFFAOYSA-N
Synonym bis cyclopentadienyl nickel ii
PubChem CID 24942185
IUPAC Name cyclopenta-1,3-diene;nickel(2+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Ni+2]

Specifications

Melting Point 171°C to 173°C
Color Green
Quantity 2 g
UN Number UN3391
Sensitivity Air and moisture sensitive
Formula Weight 188.9
Physical Form Crystalline
Chemical Name or Material Bis (cyclopentadienyl)nickel, Dry
Hazard Category H228-H302-H317-H350
Hazard Statement GHS H Statement
H250-H252-H301-H350-H317
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Toxic if swallowed.
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P240-P241-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 215-039-0
RTECSNumber QR6500000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only