missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(cyclohexanone)oxaldihydrazone, ≥99.0%, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 210°C to 214°C; 210°C to 214°C |
| CAS | 370-81-0 |
| Linear Formula | C14H22N4O2 |
| Quantity | 25 g |
| Formula Weight | 278.35 |
| Percent Purity | ≥99.0% (N); ≥99.0% |
Safety and Handling
| RTECSNumber | RO2520000 |