missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclohexanone)oxaldihydrazone, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014064
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014064 25 g
111014065 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014064 Supplier Merck Emd Millipore Supplier No. 1469025G

Cuprizone (Bis(cyclohexanone)oxaldihydrazone) is a physically unstable compound and is a well-known neurotoxic agent.

Chemical Identifiers

MDL Number MFCD00001659
Synonym Cuprizon; Cuprizone; Oxalic acid bis(cyclohexylidenehydrazide)

Specifications

Melting Point 210°C to 214°C; 210°C to 214°C
CAS 370-81-0
Linear Formula C14H22N4O2
Quantity 25 g
Formula Weight 278.35
Percent Purity ≥99.0% (N); ≥99.0%
RTECSNumber RO2520000