missing translation for 'onlineSavingsMsg'
Learn More

Bis(chloromethyl)dimethylsilane 97.0+%, TCI America™

Catalog No. B181310G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B181310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B181310G Supplier TCI America Supplier No. B181310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2917-46-6
Molecular Formula C4H10Cl2Si
Molecular Weight (g/mol) 157.109
MDL Number MFCD00000879
InChI Key TVRFAOJPBXYIRM-UHFFFAOYSA-N
PubChem CID 76212
IUPAC Name bis(chloromethyl)-dimethylsilane
SMILES C[Si](C)(CCl)CCl

Specifications

Color Red
Boiling Point 165°C
Quantity 10 g
UN Number 2920
Formula Weight 157.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(chloromethyl)dimethylsilane
EINECSNumber (2)-2065
TSCA No