missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzoylmethyl) Sulfide 99.0+%, TCI America™

Catalog No. B12655G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B12655G Supplier TCI America Supplier No. B12655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2461-80-5
Molecular Formula C16H14O2S
Molecular Weight (g/mol) 270.346
MDL Number MFCD00054497
InChI Key LHSKVFSCEMNSEC-UHFFFAOYSA-N
PubChem CID 249751
IUPAC Name 2-phenacylsulfanyl-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CSCC(=O)C2=CC=CC=C2

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 5 g
Formula Weight 270.35
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(benzoylmethyl) Sulfide