missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzonitrile)palladium(II) Dichloride 98.0+%, TCI America™

Catalog No. p-7119196
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B16681G 1 g
B16685G 5 g
2 options

Catalog No. B16681G

Supplier: TCI America B16681G

Only null left

Chemical Identifiers

CAS 14220-64-5
Molecular Formula C14H10Cl2N2Pd
Molecular Weight (g/mol) 383.57
MDL Number MFCD00013123
InChI Key QQPSWGUJSPTOSH-UHFFFAOYSA-L
Synonym bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
PubChem CID 10271322
IUPAC Name dichlorobis(phenylformonitrile)palladiumbis(ylium)
SMILES Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1

Specifications

Melting Point 130°C
Quantity 1 g
UN Number 3439
Formula Weight 383.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(benzonitrile)palladium(II) Dichloride