missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzonitrile)palladium(II) chloride, 97+%

Catalog No. p-3756909
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207900010 1 g Glass bottle
AC207900100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207900010 Supplier Thermo Scientific Chemicals Supplier No. 207900010
Only null left

CAS: 14220-64-5 | C14H10Cl2N2Pd | 383.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14220-64-5
Molecular Formula C14H10Cl2N2Pd
Molecular Weight (g/mol) 383.57
MDL Number MFCD00013123
InChI Key QQPSWGUJSPTOSH-UHFFFAOYSA-L
Synonym bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
PubChem CID 10271322
IUPAC Name benzonitrile;palladium(2+);dichloride
SMILES Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 120.0°C
Color Orange to Brown
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula (C6H5CN)2PdCl2
Quantity 1 g
Fieser 01,56; 05,31; 06,45; 09,44; 10,31; 13,34
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, toluene, chloroform
Formula Weight 383.55
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material Bis(benzonitrile)palladium(II) chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 238-085-3

RUO – Research Use Only