missing translation for 'onlineSavingsMsg'
Learn More

Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur 96.0+%, TCI America™

Catalog No. B22001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22001G 1 g
B22005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22001G Supplier TCI America Supplier No. B22001G
Only null left

Chemical Identifiers

CAS 32133-82-7
Molecular Formula C30H20F12O2S
Molecular Weight (g/mol) 672.53
MDL Number MFCD00010662
InChI Key RMIBJVUYNZSLSD-UHFFFAOYSA-N
Synonym Martin Sulfurane
PubChem CID 3608068
IUPAC Name [2-({diphenyl[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-λ⁴-sulfanyl}oxy)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzene
SMILES FC(F)(F)C(OS(OC(C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C(F)(F)F

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 672.53
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur
TSCA No
Recommended Storage Freezer