missing translation for 'onlineSavingsMsg'
Learn More

Bis(acetato)aqua[(S,S)-4,6-bis(4-isopropyl-2-oxazolin-2-yl)-m-xylene]rhodium, TCI America™

Catalog No. B419510MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B419510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B419510MG Supplier TCI America Supplier No. B419510MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 929896-28-6
Molecular Formula C24H38N2O7Rh
Molecular Weight (g/mol) 569.481
InChI Key PRWDPLSLZWMIPQ-VKBKZCJQSA-N
PubChem CID 131697923
IUPAC Name acetic acid;(4S)-2-[2,4-dimethyl-5-[(4S)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]phenyl]-4-propan-2-yl-4,5-dihydro-1,3-oxazole;rhodium;hydrate
SMILES CC1=CC(=C(C=C1C2=NC(CO2)C(C)C)C3=NC(CO3)C(C)C)C.CC(=O)O.CC(=O)O.O.[Rh]

Specifications

Quantity 10 mg
Formula Weight 566.46
Physical Form Crystalline Powder
Chemical Name or Material Bis(acetato)aqua[(S,S)-4,6-bis(4-isopropyl-2-oxazolin-2-yl)-m-xylene]rhodium