missing translation for 'onlineSavingsMsg'
Learn More

Bis(8-quinolinolato)copper(II) (purified by sublimation) 98.0+%, TCI America™

Catalog No. B38341G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38341G Supplier TCI America Supplier No. B38341G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10380-28-6
Molecular Formula C18H12CuN2O2
Molecular Weight (g/mol) 351.85
MDL Number MFCD00067392
InChI Key YXLXNENXOJSQEI-UHFFFAOYSA-L
Synonym cunilate, cuproquin, dokirin, fruitdo, quinolate, quinondo, milmer, copper oxinate, copper oxine, copper-oxine
PubChem CID 56840841
ChEBI CHEBI:82028
IUPAC Name copper(2+) bis(quinolin-8-olate)
SMILES [Cu++].[O-]C1=C2N=CC=CC2=CC=C1.[O-]C1=C2N=CC=CC2=CC=C1

Specifications

Color Black-Yellow
Quantity 1 g
UN Number 3077
Formula Weight 351.85
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(8-quinolinolato)copper(II) (purified by sublimation)
EINECSNumber (5)-0805
RTECSNumber VC5250000
TSCA Yes