missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, 99+% (metals basis)

Catalog No. AAH5518806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5518806 5 g
AAH5518803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5518806 Supplier Thermo Scientific Chemicals Supplier No. H5518806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131717-99-2 | C27H33IO3S | 564.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131717-99-2
Molecular Formula C27H33IO3S
Molecular Weight (g/mol) 564.52
MDL Number MFCD02683472
InChI Key UEJFJTOGXLEPIV-UHFFFAOYSA-M
Synonym bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis
PubChem CID 16216932
IUPAC Name bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1

Specifications

Melting Point 237°C to 240°C
Color White
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 564.52
Percent Purity ≥99%
Grade Electronic
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only