missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluorophenyl)iodonium Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. p-7124031
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52761G 1 g
B52765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52761G Supplier TCI America Supplier No. B52761G
Only null left

Chemical Identifiers

CAS 732306-64-8
Molecular Formula C13H8F5IO3S
Molecular Weight (g/mol) 466.16
MDL Number MFCD21608479
InChI Key HTFNIUDXMGKTQK-UHFFFAOYSA-M
Synonym Bis(4-fluorophenyl)iodonium Triflate
PubChem CID 15953220
IUPAC Name bis(4-fluorophenyl)iodanium;trifluoromethanesulfonate
SMILES C1=CC(=CC=C1F)[I+]C2=CC=C(C=C2)F.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 169°C
Color White
Quantity 1 g
Formula Weight 466.16
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-fluorophenyl)iodonium Trifluoromethanesulfonate