missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-fluorophenyl)iodonium Trifluoromethanesulfonate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 169°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 466.16 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(4-fluorophenyl)iodonium Trifluoromethanesulfonate |
Safety and Handling
| TSCA | No |