missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chlorobutyl) Ether 98.0+%, TCI America™

Catalog No. B092925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B092925ML 25 mL
B0929500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B092925ML Supplier TCI America Supplier No. B092925ML

Chemical Identifiers

CAS 6334-96-9
Molecular Formula C8H16Cl2O
Molecular Weight (g/mol) 199.115
MDL Number MFCD00019006
InChI Key PVBMXMKIKMJQRK-UHFFFAOYSA-N
Synonym bis 4-chlorobutyl ether, 4,4'-dichlorodibutyl ether, ether, bis 4-chlorobutyl, 4-chlorobutyl ether, di chlorobutyl ether, butane, 1,1'-oxybis 4-chloro, 1,1'-oxydi-4-chlorobutane, 1,1'-oxybis 4-chlorobutane
PubChem CID 22800
IUPAC Name 1-chloro-4-(4-chlorobutoxy)butane
SMILES C(CCCl)COCCCCCl

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 mL
Formula Weight 199.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(4-chlorobutyl) Ether
RTECSNumber KN0400000
TSCA Yes