Learn More
Bis(4-chlorobutyl) ether, 99%
CAS: 6334-96-9 | C8H16Cl2O | 199.115 g/mol
Supplier: Thermo Scientific Chemicals L1705514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Bis(4-chlorobutyl) ether | |
| 1.081 | |
| C8H16Cl2O | |
| Cl(CH2)4O(CH2)4)Cl | |
| 25 g | |
| bis 4-chlorobutyl ether, 4,4'-dichlorodibutyl ether, ether, bis 4-chlorobutyl, 4-chlorobutyl ether, di chlorobutyl ether, butane, 1,1'-oxybis 4-chloro, 1,1'-oxydi-4-chlorobutane, 1,1'-oxybis 4-chlorobutane | |
| C(CCCl)COCCCCCl | |
| 199.115 | |
| 199.12 |
| 6334-96-9 | |
| 129°C to 131°C (10 mmHg) | |
| 1.458 | |
| MFCD00019006 | |
| 1739861 | |
| PVBMXMKIKMJQRK-UHFFFAOYSA-N | |
| 1-chloro-4-(4-chlorobutoxy)butane | |
| 22800 | |
| 99% |
Chemical Identifiers
| 6334-96-9 | |
| 199.115 | |
| PVBMXMKIKMJQRK-UHFFFAOYSA-N | |
| 22800 | |
| C(CCCl)COCCCCCl |
| C8H16Cl2O | |
| MFCD00019006 | |
| bis 4-chlorobutyl ether, 4,4'-dichlorodibutyl ether, ether, bis 4-chlorobutyl, 4-chlorobutyl ether, di chlorobutyl ether, butane, 1,1'-oxybis 4-chloro, 1,1'-oxydi-4-chlorobutane, 1,1'-oxybis 4-chlorobutane | |
| 1-chloro-4-(4-chlorobutoxy)butane |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 228-713-4
RTECSNumber : KN0400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only