missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chlorobutyl) ether, 99%

Catalog No. AAL1705506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1705506 5 g
AAL1705514 25 g
AAL1705522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1705506 Supplier Thermo Scientific Chemicals Supplier No. L1705506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6334-96-9 | C8H16Cl2O | 199.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6334-96-9
Molecular Formula C8H16Cl2O
Molecular Weight (g/mol) 199.115
MDL Number MFCD00019006
InChI Key PVBMXMKIKMJQRK-UHFFFAOYSA-N
Synonym bis 4-chlorobutyl ether, 4,4'-dichlorodibutyl ether, ether, bis 4-chlorobutyl, 4-chlorobutyl ether, di chlorobutyl ether, butane, 1,1'-oxybis 4-chloro, 1,1'-oxydi-4-chlorobutane, 1,1'-oxybis 4-chlorobutane
PubChem CID 22800
IUPAC Name 1-chloro-4-(4-chlorobutoxy)butane
SMILES C(CCCl)COCCCCCl

Specifications

Density 1.081
Boiling Point 129°C to 131°C (10 mmHg)
Refractive Index 1.458
Linear Formula Cl(CH2)4O(CH2)4)Cl
Quantity 5 g
Beilstein 1739861
Formula Weight 199.12
Percent Purity 99%
Chemical Name or Material Bis(4-chlorobutyl) ether
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 228-713-4
RTECSNumber KN0400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only