missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[3-(trimethoxysilyl)propyl]amine 98.0+%, TCI America™
$53.90 - $175.20
Chemical Identifiers
| CAS | 82985-35-1 |
|---|---|
| Molecular Formula | C12H31NO6Si2 |
| Molecular Weight (g/mol) | 341.551 |
| MDL Number | MFCD00191835 |
| InChI Key | TZZGHGKTHXIOMN-UHFFFAOYSA-N |
| PubChem CID | 157882 |
| IUPAC Name | 3-trimethoxysilyl-N-(3-trimethoxysilylpropyl)propan-1-amine |
| SMILES | CO[Si](CCCNCCC[Si](OC)(OC)OC)(OC)OC |
Chemical Identifiers
| 82985-35-1 | |
| 341.551 | |
| TZZGHGKTHXIOMN-UHFFFAOYSA-N | |
| 3-trimethoxysilyl-N-(3-trimethoxysilylpropyl)propan-1-amine |
| C12H31NO6Si2 | |
| MFCD00191835 | |
| 157882 | |
| CO[Si](CCCNCCC[Si](OC)(OC)OC)(OC)OC |
Specifications
| 82985-35-1 | |
| 152°C | |
| MFCD00191835 | |
| 3082 | |
| CO[Si](CCCNCCC[Si](OC)(OC)OC)(OC)OC | |
| 341.551 | |
| 341.55 | |
| Liquid |
| Yellow | |
| C12H31NO6Si2 | |
| 25 g | |
| TZZGHGKTHXIOMN-UHFFFAOYSA-N | |
| 3-trimethoxysilyl-N-(3-trimethoxysilylpropyl)propan-1-amine | |
| 157882 | |
| ≥98.0% (T) | |
| Bis[3-(trimethoxysilyl)propyl]amine |
Safety and Handling
RTECSNumber : TX2101000
TSCA : Yes