missing translation for 'onlineSavingsMsg'
Learn More

Bis[3-(trimethoxysilyl)propyl]amine 98.0+%, TCI America™

Catalog No. B254825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B254825G 25 g
B2548100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B254825G Supplier TCI America Supplier No. B254825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82985-35-1
Molecular Formula C12H31NO6Si2
Molecular Weight (g/mol) 341.551
MDL Number MFCD00191835
InChI Key TZZGHGKTHXIOMN-UHFFFAOYSA-N
PubChem CID 157882
IUPAC Name 3-trimethoxysilyl-N-(3-trimethoxysilylpropyl)propan-1-amine
SMILES CO[Si](CCCNCCC[Si](OC)(OC)OC)(OC)OC

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 g
UN Number 3082
Formula Weight 341.55
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Bis[3-(trimethoxysilyl)propyl]amine
RTECSNumber TX2101000
TSCA Yes