missing translation for 'onlineSavingsMsg'
Learn More

Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)

Catalog No. p-7052024
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4241006 5 g
AA4241014 25 g
AA4241022 100 g
AA4241036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4241006 Supplier Thermo Scientific Chemicals Supplier No. 04241006

Coupling/cross-linking agent for precious metals | CAS: 40372-72-3 | C18H42O6S4Si2 | 538.94 g/mol

Bis[3-(triethoxysilyl)propyl]tetrasulfide is used in radial tires, rubber tire treads, carcass, tire walls, solid rubber tires and other rubber products. It acts as a chemical intermediate and as a cross linking agent. Furthermore, it acts as a coupling agent used to chemically modify silica surface. In addition, it serves as a curing agent for good heat aging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40372-72-3
Molecular Formula C18H42O6S4Si2
Molecular Weight (g/mol) 538.94
MDL Number MFCD00053751
InChI Key VTHOKNTVYKTUPI-UHFFFAOYSA-N
Synonym bis 3-triethoxysilyl propyl tetrasulfide, 4,4,15,15-tetraethoxy-3,16-dioxa-8,9,10,11-tetrathia-4,15-disilaoctadecane, unii-j98v193zry, bis 3-triethoxysilyl propyl tetrasulphide, 3,16-dioxa-8,9,10,11-tetrathia-4,15-disilaoctadecane, 4,4,15,15-tetraethoxy, silane coupler kh-858, acmc-1akw5, dsstox_cid_9362
PubChem CID 162012
IUPAC Name triethoxy-[3-(3-triethoxysilylpropyltetrasulfanyl)propyl]silane
SMILES CCO[Si](CCCSSSSCCC[Si](OCC)(OCC)OCC)(OCC)OCC

Specifications

Density 1.074 g/mL
Boiling Point 250°C (decomposition)
Flash Point 91°C (196°F)
Quantity 5 g
Solubility Information Miscible with toluene,primary alcohols,ketones,benzene,chlorinated hydrocarbons,acetonitrile,dimethylformaminde and dimethysulfoxide. Immiscible with water.
Formula Weight 538.94
Concentration or Composition (by Analyte or Components) S 22.3% (typical)
Physical Form Liquid
Chemical Name or Material Bis[3- (triethoxysilyl)propyl]tetrasulfide
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 254-896-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only