missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(3-pyridylmethyl)amine 97.0+%, TCI America™
$288.77 - $1003.02
Chemical Identifiers
| CAS | 1656-94-6 |
|---|---|
| Molecular Formula | C12H13N3 |
| Molecular Weight (g/mol) | 199.26 |
| MDL Number | MFCD00038043 |
| InChI Key | FEBQXMFOLRVSGC-UHFFFAOYSA-N |
| PubChem CID | 74257 |
| IUPAC Name | bis[(pyridin-3-yl)methyl]amine |
| SMILES | C(NCC1=CC=CN=C1)C1=CC=CN=C1 |
Chemical Identifiers
| 1656-94-6 | |
| 199.26 | |
| FEBQXMFOLRVSGC-UHFFFAOYSA-N | |
| bis[(pyridin-3-yl)methyl]amine |
| C12H13N3 | |
| MFCD00038043 | |
| 74257 | |
| C(NCC1=CC=CN=C1)C1=CC=CN=C1 |
Specifications
| 1656-94-6 | |
| C12H13N3 | |
| 5 mL | |
| FEBQXMFOLRVSGC-UHFFFAOYSA-N | |
| bis[(pyridin-3-yl)methyl]amine | |
| 74257 | |
| ≥97.0% (GC) | |
| Bis(3-pyridylmethyl)amine |
| Yellow | |
| MFCD00038043 | |
| 3,3′C-Dipicolylamine, alpha,alpha′C-Iminodi(3-picoline) | |
| C(NCC1=CC=CN=C1)C1=CC=CN=C1 | |
| 199.26 | |
| 199.26 | |
| Liquid |
Safety and Handling
RTECSNumber : UT4397000
TSCA : No