missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-pyridylmethyl)amine 97.0+%, TCI America™

Catalog No. D11995ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11995ML 5 mL
D119925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11995ML Supplier TCI America Supplier No. D11995ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1656-94-6
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.26
MDL Number MFCD00038043
InChI Key FEBQXMFOLRVSGC-UHFFFAOYSA-N
PubChem CID 74257
IUPAC Name bis[(pyridin-3-yl)methyl]amine
SMILES C(NCC1=CC=CN=C1)C1=CC=CN=C1

Specifications

Color Yellow
Quantity 5 mL
Synonym 3,3′C-Dipicolylamine, alpha,alpha′C-Iminodi(3-picoline)
Formula Weight 199.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(3-pyridylmethyl)amine
RTECSNumber UT4397000
TSCA No