missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-methyl-1-phenyl-5-pyrazolone) 97.0+%, TCI America™

Catalog No. p-7118487
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
B050825G 25 g
B0508250G 250 g
2 options

Catalog No. B050825G

Supplier: TCI America B050825G

Only null left

Chemical Identifiers

CAS 7477-67-0
Molecular Formula C20H18N4O2
Molecular Weight (g/mol) 346.39
MDL Number MFCD00020765
InChI Key FORCWSNQDMPPOC-UHFFFAOYSA-N
Synonym bispyrazolone, 4,4'-bis 1-phenyl-3-methyl-5-pyrazolone, 4,4'-bi-3h-pyrazole-3,3'-dione, 2,2',4,4'-tetrahydro-5,5'-dimethyl-2,2'-diphenyl, 4,4'-bis 3-methyl-1-phenyl-5-pyrazolone, 4,4'-bi-2-pyrazoline-5,5'-dione, 3,3'-dimethyl-1,1'-diphenyl, 3,3'-dimethyl-1,1'-diphenyl-1h,1'h-4,4'-bipyrazole-5,5' 4h,4'h-dione, 5-methyl-4-3-methyl-5-oxo-1-phenyl-4h-pyrazol-4-yl-2-phenyl-4h-pyrazol-3-one, 4,5'-dione, 3,3'-dimethyl-1,1'-diphenyl
PubChem CID 95807
IUPAC Name 5-methyl-4-(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)-2-phenyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1C2C(=NN(C2=O)C3=CC=CC=C3)C)C4=CC=CC=C4

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 346.39
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-methyl-1-phenyl-5-pyrazolone) [for Determination of CN, Vitamin B12, etc]