missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 97%

Catalog No. AAH6196406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6196406 5 g
AAH6196403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6196406 Supplier Thermo Scientific Chemicals Supplier No. H6196406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 811862-77-8 | C42H44O10Pd | 815.22 g/mol

Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(3,5,3′,5′-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 811862-77-8
Molecular Formula C42H44O10Pd
Molecular Weight (g/mol) 815.22
MDL Number MFCD07369799
InChI Key RTGAJOPJZJDWAX-UHFFFAOYSA-N
Synonym bis 3,5,3',5'-dimethoxydibenzylideneacetone palladium 0, pd dmdba 2, bis 353'5'-dimethoxydibenzylideneace, bis 3,5,3',5'-dimethoxydibenzylideneacetone palladium
PubChem CID 24777333
SMILES [Pd].COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1.COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1

Specifications

Melting Point 170°C to 178°C
Color Red
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name 1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one;palladium
Formula Weight 815.23
Concentration or Composition (by Analyte or Components) Pd 13%
Physical Form Powder
Chemical Name or Material Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only