missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-pyridylmethyl)amine 98.0+%, TCI America™

Catalog No. D22285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22285G 5 g
D222825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22285G Supplier TCI America Supplier No. D22285G
Only null left

Chemical Identifiers

CAS 1539-42-0
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD00129044
InChI Key KXZQYLBVMZGIKC-UHFFFAOYSA-N
Synonym 2,2′C-Dipicolylamine, alpha,alpha′C-Iminodi(2-picoline)
PubChem CID 73759
IUPAC Name 1-pyridin-2-yl-N-(pyridin-2-ylmethyl)methanamine
SMILES C1=CC=NC(=C1)CNCC2=CC=CC=N2

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 g
Formula Weight 199.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Bis(2-pyridylmethyl)amine