missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-oxo-3-oxazolidinyl)phosphinic Chloride 97.0+%, TCI America™

Catalog No. p-7118926
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12135G 5 g
B121325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12135G Supplier TCI America Supplier No. B12135G
Only null left

Chemical Identifiers

CAS 68641-49-6
Molecular Formula C6H8ClN2O5P
Molecular Weight (g/mol) 254.563
MDL Number MFCD00010077
InChI Key KLDLRDSRCMJKGM-UHFFFAOYSA-N
Synonym bis 2-oxo-3-oxazolidinyl phosphinic chloride, bop-cl, bis 2-oxooxazolidin-3-yl phosphinic chloride, ccris 2607, phosphinic chloride, bis 2-oxo-3-oxazolidinyl, bis 2-oxo-3-oxazolidinyl phosphonic chloride, bis 2-oxo-1,3-oxazolidin-3-yl phosphinoyl chloride, bis-2-oxo-3-oxazolidinyl phosphinic chloride, n,n-bis 2-oxo-3-oxazolidinyl phosphinic chloride, bis 2-oxo-3-oxazolidinyl phosphinic chloride bop-cl
PubChem CID 152842
IUPAC Name 3-[chloro-(2-oxo-1,3-oxazolidin-3-yl)phosphoryl]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1P(=O)(N2CCOC2=O)Cl

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 254.56
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-oxo-3-oxazolidinyl)phosphinic Chloride
RTECSNumber SZ5871000
TSCA No