missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%

Catalog No. AC310060050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310060050 5 g Glass bottle
AC310060250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310060050 Supplier Thermo Scientific Chemicals Supplier No. 310060050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 68641-49-6 | C6H8ClN2O5P | 254.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68641-49-6
Molecular Formula C6H8ClN2O5P
Molecular Weight (g/mol) 254.57
MDL Number MFCD00010077
InChI Key KLDLRDSRCMJKGM-UHFFFAOYSA-N
Synonym bis 2-oxo-3-oxazolidinyl phosphinic chloride, bop-cl, bis 2-oxooxazolidin-3-yl phosphinic chloride, ccris 2607, phosphinic chloride, bis 2-oxo-3-oxazolidinyl, bis 2-oxo-3-oxazolidinyl phosphonic chloride, bis 2-oxo-1,3-oxazolidin-3-yl phosphinoyl chloride, bis-2-oxo-3-oxazolidinyl phosphinic chloride, n,n-bis 2-oxo-3-oxazolidinyl phosphinic chloride, bis 2-oxo-3-oxazolidinyl phosphinic chloride bop-cl
PubChem CID 152842
IUPAC Name 3-[chloro-(2-oxo-1,3-oxazolidin-3-yl)phosphoryl]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1P(=O)(N2CCOC2=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 191°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 254.57
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only