missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%

Catalog No. AAL0877514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0877514 25 g
AAL0877506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0877514 Supplier Thermo Scientific Chemicals Supplier No. L0877514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68641-49-6 | C6H8ClN2O5P | 254.563 g/mol

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 68641-49-6
Molecular Formula C6H8ClN2O5P
Molecular Weight (g/mol) 254.563
MDL Number MFCD00010077
InChI Key KLDLRDSRCMJKGM-UHFFFAOYSA-N
Synonym bis 2-oxo-3-oxazolidinyl phosphinic chloride, bop-cl, bis 2-oxooxazolidin-3-yl phosphinic chloride, ccris 2607, phosphinic chloride, bis 2-oxo-3-oxazolidinyl, bis 2-oxo-3-oxazolidinyl phosphonic chloride, bis 2-oxo-1,3-oxazolidin-3-yl phosphinoyl chloride, bis-2-oxo-3-oxazolidinyl phosphinic chloride, n,n-bis 2-oxo-3-oxazolidinyl phosphinic chloride, bis 2-oxo-3-oxazolidinyl phosphinic chloride bop-cl
PubChem CID 152842
IUPAC Name 3-[chloro-(2-oxo-1,3-oxazolidin-3-yl)phosphoryl]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1P(=O)(N2CCOC2=O)Cl

Specifications

Melting Point ∽195°C (decomposition)
Quantity 25 g
UN Number UN3261
Beilstein 3654596
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 254.57
Percent Purity 97%
Chemical Name or Material Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber SZ5871000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only