missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-morpholinoethyl) Ether 85.0+%, TCI America™

Catalog No. p-7119260
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B178425G 25 g
B1784500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B178425G Supplier TCI America Supplier No. B178425G

Chemical Identifiers

CAS 6425-39-4
Molecular Formula C12H24N2O3
Molecular Weight (g/mol) 244.335
MDL Number MFCD00072740
InChI Key ZMSQJSMSLXVTKN-UHFFFAOYSA-N
PubChem CID 80900
IUPAC Name 4-[2-(2-morpholin-4-ylethoxy)ethyl]morpholine
SMILES C1COCCN1CCOCCN2CCOCC2

Specifications

Melting Point -28°C
Boiling Point 192°C
Quantity 25 g
Synonym 4,4′C-(3-Oxapentane-1,5-diyl)bismorpholine
Formula Weight 244.34
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-morpholinoethyl) Ether
EINECSNumber (5)-6265
TSCA Yes