missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phtalate de Bis(2-méthoxyéthyle) 96,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 340°C |
| Quantity | 500 g |
| Formula Weight | 282.29 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Bis(2-methoxyethyl) Phthalate |
Sécurité et manipulation
| EINECSNumber | (3)-2962 |
| RTECSNumber | TI1400000 |
| TSCA | Yes |