missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) Phthalate 96.0+%, TCI America™

Catalog No. P096525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P096525G 25 g
P0965500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P096525G Supplier TCI America Supplier No. P096525G

Chemical Identifiers

CAS 117-82-8
Molecular Formula C14H18O6
Molecular Weight (g/mol) 282.29
MDL Number MFCD00042842
InChI Key HSUIVCLOAAJSRE-UHFFFAOYSA-N
Synonym bis 2-methoxyethyl phthalate, kesscoflex mcp, 2-methoxyethyl phthalate, dmep, bis methoxyethyl phthalate, di 2-methoxyethyl phthalate, kodaflex dmep, methyl glycol phthalate, dimethoxyethyl phthalate, dimethylglycol phthalate
PubChem CID 8344
IUPAC Name 1,2-bis(2-methoxyethyl) benzene-1,2-dicarboxylate
SMILES COCCOC(=O)C1=CC=CC=C1C(=O)OCCOC

Specifications

Color Colorless
Boiling Point 340°C
Quantity 25 g
Formula Weight 282.29
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-methoxyethyl) Phthalate
EINECSNumber (3)-2962
RTECSNumber TI1400000
TSCA Yes