missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene
CAS: 202289-38-1 | C6H14F3NO2S | 221.24 g/mol
$129.43 - $1235.17
Specifications
| Chemical Name or Material | [Bis(2-methoxyethyl)amino]sulfur trifluoride |
|---|---|
| CAS | 202289-38-1,108-88-3 |
| CAS Min % | 49.0,49.0 |
| CAS Max % | 51.0,51.0 |
| Color | Orange-Yellow |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC368960050
|
Thermo Scientific Chemicals
368960050 |
5 g | Plastic bottle |
Each for $129.43
|
|
||||
|
AC368960250
|
Thermo Scientific Chemicals
368960250 |
25 g | Plastic bottle |
Each for $421.54
|
|
||||
|
AC368961000
|
Thermo Scientific Chemicals
368961000 |
100 g | Plastic bottle |
Each for $1,235.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
| [Bis(2-methoxyethyl)amino]sulfur trifluoride | |
| 49.0,49.0 | |
| Orange-Yellow | |
| 4°C | |
| C6H14F3NO2S | |
| 15,2899 | |
| bis 2-methoxyethyl aminosulfur trifluoride, deoxo-fluor, 2-methoxy-n-2-methoxyethyl-n-trifluorothio-ethanamine, deoxo-fluor r, bis 2-methoxyethyl aminosulphurtrifluoride, bast, bis 2-methoxyethyl amino sulfur trifluoride, bis 2-methoxyethyl aminosulphur trifluoride | |
| COCCN(CCOC)S(F)(F)F | |
| 221.24 | |
| 221.24 | |
| Liquid |
| 202289-38-1,108-88-3 | |
| 51.0,51.0 | |
| 1.2000g/mL | |
| Plastic bottle | |
| (CH3OCHCH2)2NSF3 | |
| 1.2 | |
| APOYTRAZFJURPB-UHFFFAOYSA-N | |
| 2-methoxy-N-(2-methoxyethyl)-N-(trifluoro-$l^{4}-sulfanyl)ethanamine | |
| 2734690 | |
| ≥95% (NMR, neat substance) |
RUO – Research Use Only