missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) Adipate 98.0+%, TCI America™

Catalog No. p-7116750
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016525ML 25 mL
A0165500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A016525ML Supplier TCI America Supplier No. A016525ML
Only null left

Chemical Identifiers

CAS 106-00-3
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.302
MDL Number MFCD00041905
InChI Key GVRNUDCCYWKHMV-UHFFFAOYSA-N
Synonym bis 2-methoxyethyl adipate, hexanedioic acid, bis 2-methoxyethyl ester, bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate, adipic acid, bis 2-methoxyethyl ester, adipic acid bis 2-methoxyethyl ester, hexanedioic acid, 1,6-bis 2-methoxyethyl ester, 1,6-bis 2-methoxyethyl hexanedioate
PubChem CID 66046
IUPAC Name bis(2-methoxyethyl) hexanedioate
SMILES COCCOC(=O)CCCCC(=O)OCCOC

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 mL
Formula Weight 262.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-methoxyethyl) Adipate
EINECSNumber (2)-0883
TSCA Yes