missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl)dithiocarbamic Acid Zinc(II) Salt 98.0+%, TCI America™

$109.98 - $344.86

Chemical Identifiers

CAS 19163-92-9
Molecular Formula C10H20N2O4S4Zn
Molecular Weight (g/mol) 425.9
MDL Number MFCD00070508
InChI Key BRJVMKALQCUXPN-UHFFFAOYSA-L
Synonym Bis[bis(2-hydroxyethyl)dithiocarbamato]zinc(II)
PubChem CID 87948
IUPAC Name zinc;N,N-bis(2-hydroxyethyl)carbamodithioate
SMILES C(CO)N(CCO)C(=S)[S-].C(CO)N(CCO)C(=S)[S-].[Zn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B04925G
View Documents
TCI America
B04925G
5 g N/A
Only null left
 
B049225G
View Documents
TCI America
B049225G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

19163-92-9
425.9
BRJVMKALQCUXPN-UHFFFAOYSA-L
87948
C(CO)N(CCO)C(=S)[S-].C(CO)N(CCO)C(=S)[S-].[Zn+2]
C10H20N2O4S4Zn
MFCD00070508
Bis[bis(2-hydroxyethyl)dithiocarbamato]zinc(II)
zinc;N,N-bis(2-hydroxyethyl)carbamodithioate

Specifications

19163-92-9
White-Yellow
MFCD00070508
Bis[bis(2-hydroxyethyl)dithiocarbamato]zinc(II)
C(CO)N(CCO)C(=S)[S-].C(CO)N(CCO)C(=S)[S-].[Zn+2]
425.9
425.90
Crystalline Powder
151°C
C10H20N2O4S4Zn
5 g
BRJVMKALQCUXPN-UHFFFAOYSA-L
zinc;N,N-bis(2-hydroxyethyl)carbamodithioate
87948
≥98.0% (T)
Bis(2-hydroxyethyl)dithiocarbamic Acid Zinc(II) Salt
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'