missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-hydroxyethyl)dithiocarbamic Acid Zinc(II) Salt 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 151°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 425.90 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(2-hydroxyethyl)dithiocarbamic Acid Zinc(II) Salt |
Safety and Handling
| TSCA | No |