missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane 99.0+%, TCI America™

Catalog No. p-7119081
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
B149325G 25 g
B1493250G 250 g
2 options

Catalog No. B149325G

Supplier: TCI America B149325G

Only null left

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Color White
Quantity 25 g
Formula Weight 209.24
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane [Good′s buffer component for biological res