missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) sebacate, 97%

Catalog No. AC269920010
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269920010 1 L Glass bottle
AC269920025 2.5 L Glass bottle
AC269920100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC269920010 Supplier Thermo Scientific Chemicals Supplier No. 269920010

CAS: 122-62-3 | C26H50O4 | 426.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-62-3
Molecular Formula C26H50O4
Molecular Weight (g/mol) 426.68
MDL Number MFCD00009497
InChI Key VJHINFRRDQUWOJ-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate
PubChem CID 31218
IUPAC Name bis(2-ethylhexyl) decanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

Specifications

Acidity 0.15mg KOH/g max.
Melting Point -55°C
Color Yellow
Density 0.9100g/mL
Boiling Point 212°C (1.0 mmHg)
Flash Point 215°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4500 to 1.455
Quantity 1 L
Saponification Value 260 to 265mg KOH/g
Beilstein 02,181
Residue after Evaporation 12% (10g, 6 hrs, 200°C) max.
Specific Gravity 0.91
Merck Index 15,1255
Solubility Information Solubility in water: insoluble
Viscosity ca 23 mPa.s (20°C)
Formula Weight 426.67
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) sebacate
Show More Show Less
EINECSNumber 204-558-8

RUO – Research Use Only