missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) sebacate, 95%

CAS: 122-62-3 | C26H50O4 | 426.68 g/mol

$66.37 - $529.59

Chemical Identifiers

CAS 122-62-3
Molecular Formula C26H50O4
Molecular Weight (g/mol) 426.68
MDL Number MFCD00009497
InChI Key VJHINFRRDQUWOJ-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate
PubChem CID 31218
IUPAC Name bis(2-ethylhexyl) decanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA16049AK
View Documents
Thermo Scientific Chemicals
A16049AK
250 mL N/A
Only null left
 
AAA16049AU
View Documents
Thermo Scientific Chemicals
A16049AU
1000 mL N/A
Only null left
 
AAA160490J
View Documents
Thermo Scientific Chemicals
A160490J
5000 mL N/A
Only null left
 
Description

Description

Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

122-62-3
426.68
VJHINFRRDQUWOJ-UHFFFAOYNA-N
31218
CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
C26H50O4
MFCD00009497
bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate
bis(2-ethylhexyl) decanedioate

Specifications

122-62-3
211°C to 213°C (1 mmHg)
C26H50O4
MFCD00009497
14,1251
Soluble in water (0.1g/L at 20°C).
CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
426.68
426.68
Bis(2-ethylhexyl) sebacate
0.914
>110°C (230°F)
1.451
250 mL
bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate
VJHINFRRDQUWOJ-UHFFFAOYNA-N
bis(2-ethylhexyl) decanedioate
31218
95%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-558-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only