missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Hydrogen Phosphate 95.0+%, TCI America™
GREENER_CHOICE

Catalog No. P0811500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0811500ML 500 mL
P081125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0811500ML Supplier TCI America Supplier No. P0811500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 298-07-7
Molecular Formula C16H35O4P
Molecular Weight (g/mol) 322.43
MDL Number MFCD00009492
InChI Key SEGLCEQVOFDUPX-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl hydrogen phosphate, bis 2-ethylhexyl phosphate, hdehp, di 2-ethylhexyl phosphate, dehpa extractant, di 2-ethylhexyl phosphoric acid, escaid 100, d 2ehpa, bis 2-ethylhexyl phosphoric acid
PubChem CID 9275
IUPAC Name bis[(2-ethylhexyl)oxy]phosphinic acid
SMILES CCCCC(CC)COP(O)(=O)OCC(CC)CCCC

Specifications

Melting Point -50°C
Color Yellow
Quantity 500 mL
UN Number 1902
Formula Weight 322.43
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Hydrogen Phosphate [for Rare Metals Extraction]
EINECSNumber (2)-1986
RTECSNumber TB7875000
TSCA Yes