missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Adipate 98.0+%, TCI America™

Catalog No. A016325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016325ML 25 mL
A0163500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A016325ML Supplier TCI America Supplier No. A016325ML

Chemical Identifiers

CAS 103-23-1
Molecular Formula C22H42O4
Molecular Weight (g/mol) 370.574
MDL Number MFCD00009496
InChI Key SAOKZLXYCUGLFA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
PubChem CID 7641
ChEBI CHEBI:34675
IUPAC Name bis(2-ethylhexyl) hexanedioate
SMILES CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 214°C
Quantity 25 mL
Formula Weight 370.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AU9700000
TSCA Yes