missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) adipate, 99%

Catalog No. AAL1019514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1019514 25 g
AAL1019522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1019514 Supplier Thermo Scientific Chemicals Supplier No. L1019514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-23-1 | C22H42O4 | 370.574 g/mol

It is employed as a plasticizer. It is a skin conditioning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a plasticizer. It is a skin conditioning solvent.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 103-23-1
Molecular Formula C22H42O4
Molecular Weight (g/mol) 370.574
MDL Number MFCD00009496
InChI Key SAOKZLXYCUGLFA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
PubChem CID 7641
ChEBI CHEBI:34675
IUPAC Name bis(2-ethylhexyl) hexanedioate
SMILES CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

Specifications

Melting Point -67°C
Density 0.99
Boiling Point 166°C to 168°C (1 mmHg)
Flash Point 196°C (384°F)
Refractive Index 1.447
Quantity 25 g
Beilstein 1803774
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 370.58
Percent Purity 99%
Chemical Name or Material Bis(2-ethylhexyl) adipate
Show More Show Less
Hazard Category H373
Precautionary Statement P260-P314-P501c
EINECSNumber 203-090-1
RTECSNumber AU9700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only