missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) adipate, 99%, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | <-70°C |
| CAS | 103-23-1 |
| Density | 0.925 g/mL (at 20°C) |
| Boiling Point | 175°C (2 mmHg) |
| Flash Point | 196°C |
| Packaging | Glass Bottle |
| Linear Formula | [-CH2 CH2 CO2 CH2 CH(C2H5)(CH2)3 CH3]2 |
| Quantity | 5 mL |
| Formula Weight | 370.57 |
| Percent Purity | 99% |