missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) 4-Cyclohexene-1,2-dicarboxylate 97.0+%, TCI America™

Catalog No. T060525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T060525G 25 g
T0605500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T060525G Supplier TCI America Supplier No. T060525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2915-49-3
Molecular Formula C24H42O4
Molecular Weight (g/mol) 394.596
MDL Number MFCD00059565
InChI Key SVVBLKNHJWTATO-UHFFFAOYSA-N
Synonym 4-Cyclohexene-1,2-dicarboxylic Acid Di(2-ethylhexyl) Ester, Dioctyl 4-Cyclohexene-1,2-dicarboxylate, 4-Cyclohexene-1,2-dicarboxylic Acid Bis(2-ethylhexyl) Ester
PubChem CID 520413
IUPAC Name bis(2-ethylhexyl) cyclohex-4-ene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1CC=CCC1C(=O)OCC(CC)CCCC

Specifications

Color Colorless
Quantity 25 g
Formula Weight 394.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) 4-Cyclohexene-1,2-dicarboxylate
EINECSNumber (3)-2437
TSCA No