missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethoxyethyl)amine 98.0+%, TCI America™

Catalog No. B136425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B136425ML 25 mL
B1364500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B136425ML Supplier TCI America Supplier No. B136425ML

Chemical Identifiers

CAS 124-21-0
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.25
MDL Number MFCD00039904
InChI Key QDZOFZFDBDYWJX-UHFFFAOYSA-N
PubChem CID 547396
IUPAC Name bis(2-ethoxyethyl)amine
SMILES CCOCCNCCOCC

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 mL
UN Number 2735
Synonym 2,2′C-Diethoxydiethylamine
Formula Weight 161.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethoxyethyl)amine