missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-dimethylaminoethyl) Ether 98.0+%, TCI America™

Catalog No. p-7118974
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B129125ML 25 mL
B1291500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B129125ML Supplier TCI America Supplier No. B129125ML
Only null left

Chemical Identifiers

CAS 3033-62-3
Molecular Formula C8H20N2O
Molecular Weight (g/mol) 160.26
MDL Number MFCD00059199
InChI Key GTEXIOINCJRBIO-UHFFFAOYSA-N
Synonym bis 2-dimethylaminoethyl ether, niax catalyst al, kalpur pc, toyocat ets, niax a 1, texacat zf 20, dabco bl 19i, dabco bl 11, dabco bl 19
PubChem CID 18204
IUPAC Name {2-[2-(dimethylamino)ethoxy]ethyl}dimethylamine
SMILES CN(C)CCOCCN(C)C

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 mL
UN Number 2929
Formula Weight 160.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-dimethylaminoethyl) Ether
EINECSNumber (2)-0390
RTECSNumber KR9460000
TSCA Yes