missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-dimethylaminoethyl) ether, 98%

CAS: 3033-62-3 | C8H20N2O | 160.26 g/mol

$263.03 - $263.03

Chemical Identifiers

CAS 3033-62-3
Molecular Formula C8H20N2O
Molecular Weight (g/mol) 160.26
MDL Number MFCD00059199
InChI Key GTEXIOINCJRBIO-UHFFFAOYSA-N
Synonym bis 2-dimethylaminoethyl ether, niax catalyst al, kalpur pc, toyocat ets, niax a 1, texacat zf 20, dabco bl 19i, dabco bl 11, dabco bl 19
PubChem CID 18204
IUPAC Name 2-[2-(dimethylamino)ethoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOCCN(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0988418
View Documents
Thermo Scientific Chemicals
L0988418
50 g
Each for $263.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3033-62-3
160.26
GTEXIOINCJRBIO-UHFFFAOYSA-N
18204
CN(C)CCOCCN(C)C
C8H20N2O
MFCD00059199
bis 2-dimethylaminoethyl ether, niax catalyst al, kalpur pc, toyocat ets, niax a 1, texacat zf 20, dabco bl 19i, dabco bl 11, dabco bl 19
2-[2-(dimethylamino)ethoxy]-N,N-dimethylethanamine

Specifications

3033-62-3
179°C to 180°C
C8H20N2O
MFCD00059199
UN2922
bis 2-dimethylaminoethyl ether, niax catalyst al, kalpur pc, toyocat ets, niax a 1, texacat zf 20, dabco bl 19i, dabco bl 11, dabco bl 19
CN(C)CCOCCN(C)C
160.26
160.26
Bis(2-dimethylaminoethyl) ether
0.85
66°C (151°F)
1.43
50 g
1739668
GTEXIOINCJRBIO-UHFFFAOYSA-N
2-[2-(dimethylamino)ethoxy]-N,N-dimethylethanamine
18204
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H331-H314-H318-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H332-H311-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 221-220-5

RTECSNumber : KR9460000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only