missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-dimethylaminoethyl) ether, 98%

Catalog No. AAL0988418
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0988418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0988418 Supplier Thermo Scientific Chemicals Supplier No. L0988418
Only null left

CAS: 3033-62-3 | C8H20N2O | 160.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3033-62-3
Molecular Formula C8H20N2O
Molecular Weight (g/mol) 160.26
MDL Number MFCD00059199
InChI Key GTEXIOINCJRBIO-UHFFFAOYSA-N
Synonym bis 2-dimethylaminoethyl ether, niax catalyst al, kalpur pc, toyocat ets, niax a 1, texacat zf 20, dabco bl 19i, dabco bl 11, dabco bl 19
PubChem CID 18204
IUPAC Name 2-[2-(dimethylamino)ethoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOCCN(C)C

Specifications

Density 0.85
Boiling Point 179°C to 180°C
Flash Point 66°C (151°F)
Refractive Index 1.43
Quantity 50 g
UN Number UN2922
Beilstein 1739668
Formula Weight 160.26
Percent Purity 98%
Chemical Name or Material Bis(2-dimethylaminoethyl) ether
Hazard Category H227-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H311-H331-H314-H318-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 221-220-5
RTECSNumber KR9460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only