missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-dimethylaminoethyl) Disulfide Dihydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 281.30 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(2-dimethylaminoethyl) Disulfide Dihydrochloride |
Safety and Handling
| EINECSNumber | (2)-3391 |
| RTECSNumber | KR7300000 |
| TSCA | Yes |